MilliporeSigma
  • Topological polar surface area defines substrate transport by multidrug resistance associated protein 1 (MRP1/ABCC1).
Topological polar surface area defines substrate transport by multidrug resistance associated protein 1 (MRP1/ABCC1).
Journal of medicinal chemistry (2009-02-06)
Janaina Fernandes, Cerli R Gattass
ABSTRACT
Multidrug resistance-associated protein 1 (MRP1/ABCC1) is a very promiscuous transporter. Herein we used topological polar surface area (TPSA), a descriptor defined as the sum of surfaces of polar atoms in a molecule, to analyze drug transport by MRP1. We suggested that compounds with high TPSA are transported while those with low TPSA are not. The conjugation to GSH increases TPSA values favoring transport. A strong correlation between TPSA and transport properties (K(m)) was also found.
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione oxidized, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione oxidized, lyophilized powder
Sigma-Aldrich
L-Glutathione oxidized, BioXtra, ≥98%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%