MilliporeSigma
  • Tanshinone II is a potent candidate for treatment of lipopolysaccharide-induced acute lung injury in rat model.
Tanshinone II is a potent candidate for treatment of lipopolysaccharide-induced acute lung injury in rat model.
Oncology letters (2018-02-13)
Ling Li, Yong-Gang Zhang, Yu-Fei Tan, Jing-Jing Zhao, Hua-Ru Zhang, Bing Zhao
ABSTRACT
The present study aimed to investigate the effect of tanshinone II, isolated from Salvia miltiorrhiza Bunge, on lipopolysaccharide (LPS)-induced acute lung injury (ALI) in rats. Male Sprague-Dawley rats were divided into three groups: Control, LPS and tanshinone II. Animals in the tanshinone II and LPS groups were administered 10 mg/kg LPS, whereas those in the control group received an equal volume of normal saline. Tanshinone II treatment group were injected with 30 nm/kg tanshinone II at 1 h after LPS administration. The results revealed that LPS administration increased the bronchoalveolar lavage fluid protein concentration significantly compared with the control group. However, tanshinone II treatment significantly inhibited the LPS-induced increase in protein level. Treatment of the LPS-administered rats with tanshinone II prevented the formation of pulmonary edema, which was evidenced by low values for wet to dry lung weight ratio. The activity of myeloperoxidase and expression of malondialdehyde were significantly lower in lung homogenates from the tanshinone II group compared with the LPS group. Furthermore, tanshinone II treatment inhibited the expression of tumor necrosis factor-α and interleukin-6 in the blood plasma. Tissue sections of the tanshinone II group exhibited normal morphology and absence of neutrophil accumulation. However, in the LPS group, neutrophils accumulated and penetrated into the pulmonary tissues. These results suggested that tanshinone II protects the rats from LPS-induced ALI. Therefore tanshinone II may have clinical applications in the treatment of ALI.
Sigma-Aldrich
Rat IL-6 ELISA Kit, for serum, plasma and cell culture supernatants