MilliporeSigma
  • MCM2 and Carbonic Anhydrase 9 Are Novel Potential Targets for Neuroblastoma Pharmacological Treatment.
MCM2 and Carbonic Anhydrase 9 Are Novel Potential Targets for Neuroblastoma Pharmacological Treatment.
Biomedicines (2020-11-07)
Patrizia Garbati, Raffaella Barbieri, Davide Cangelosi, Carlo Zanon, Delfina Costa, Alessandra Eva, Stefano Thellung, Matilde Calderoni, Francesca Baldini, Gian Paolo Tonini, Paola Modesto, Tullio Florio, Aldo Pagano
ABSTRACT
To overcome the lack of effective pharmacological treatments for high-risk neuroblastoma (HR-NB), the development of novel in vitro and in vivo models that better recapitulate the disease is required. Here, we used an in vitro multiclonal cell model encompassing NB cell differentiation stages, to identify potential novel pharmacological targets. This model allowed us to identify, by low-density RT-PCR arrays, two gene sets, one over-expressed during NB cell differentiation, and the other up-regulated in more malignant cells. Challenging two HR-NB gene expression datasets, we found that these two gene sets are related to high and low survival, respectively. Using mouse NB cisplatin-treated xenografts, we identified two genes within the list associated to the malignant stage (MCM2 and carbonic anhydrase 9), whose expression is positively correlated with tumor growth. Thus, we tested their pharmacological targeting as potential therapeutic strategy. We measured mice survival and tumor growth rate after xenografts of human NB treated with cisplatin in the presence of MCM2/carbonic anhydrase 9 inhibitors (ciprofloxacin and acetazolamide). MCM2 or carbonic anhydrase 9 inhibition significantly increased cisplatin activity, supporting their possible testing for NB therapy.
Sigma-Aldrich
Anti-Disialoganglioside GD2 Antibody, clone 14G2a, clone 14.2Ga, Chemicon®, from mouse
Sigma-Aldrich
Anti-Nestin Antibody, clone 10C2, clone 10C2, Chemicon®, from mouse