MilliporeSigma
  • Calculated cell-specific intracellular hydrogen peroxide concentration: Relevance in cancer cell susceptibility during ascorbate therapy.
Calculated cell-specific intracellular hydrogen peroxide concentration: Relevance in cancer cell susceptibility during ascorbate therapy.
Free radical biology & medicine (2018-03-31)
Dieanira Erudaitius, Jacqueline Mantooth, Andrew Huang, Jesse Soliman, Claire M Doskey, Garry R Buettner, Victor G J Rodgers
ABSTRACT
The high extracellular hydrogen peroxide (H2O2) concentrations generated during pharmacological ascorbate (P-AscH-) therapy has been shown to exhibit a high flux into susceptible cancer cells leading to a decrease in clonogenic survival. It is hypothesized that the intracellular H2O2 concentration for susceptibility is independent of cell type and that the variation observed in dosing is associated with differences in the cell-specific overall steady-state intracellular H2O2 concentration values. The steady-state variation in intracellular H2O2 concentration is coupled to a number of cellular specific transport and reaction factors including catalase activity and membrane permeability. Here a lumped-parameter mathematical modeling approach, assuming a catalase-dominant peroxide removal mechanism, is used to calculate intracellular H2O2 concentration for several cell lines. Experimental measurements of critical parameters pertaining to the model are obtained. The cell lines investigated are normal pancreatic cells, H6c7, the pancreatic cancer cell line, MIA PaCa-2 and the glioblastoma cell lines, LN-229, T98G, and U-87; all which vary in susceptibility. The intracellular H2O2 concentration estimates are correlated with the clonogenic surviving fraction for each cell line, in-vitro. The results showed that, despite the fact that the experimental parameters including catalase concentration and plasma membrane permeability demonstrated significant variability across cell lines, the calculated steady-state intracellular to extracellular H2O2 concentration ratio did not vary significantly across cell lines. Thus, the calculated intracellular H2O2 concentration is not unique in characterizing susceptibility. These results imply that, although intracellular H2O2 concentration plays a key role in cellular susceptibility to P-AscH- adjuvant therapy, its overall contribution in a unifying mechanism across cell types is complex.
Sigma-Aldrich
Peroxisome Isolation Kit, isolate peroxisomes from tissues and cells