MilliporeSigma
  • Glyphosate exposures and kidney injury biomarkers in infants and young children.
Glyphosate exposures and kidney injury biomarkers in infants and young children.
Environmental pollution (Barking, Essex : 1987) (2019-11-05)
Leonardo Trasande, Sandra India Aldana, Howard Trachtman, Kurunthachalam Kannan, Deborah Morrison, Dimitri A Christakis, Kathryn Whitlock, Mary Jo Messito, Rachel S Gross, Rajendiran Karthikraj, Sheela Sathyanarayana
ABSTRACT
The goal of this study was to assess biomarkers of exposure to glyphosate and assess potential associations with renal function in children. Glyphosate is used ubiquitously in agriculture worldwide. While previous studies have indicated that glyphosate may have nephrotoxic effects, few have examined potential effects on kidney function in children. We leveraged three cohorts across different phases of child development and measured urinary levels of glyphosate. We evaluated associations of glyphosate with three biomarkers of kidney injury: albuminuria (ACR), neutrophil gelatinase-associated lipocalin (NGAL), and kidney injury marker 1 (KIM-1). Multivariable regression analyses examined associations of glyphosate with kidney injury biomarkers controlling for covariates. We identified glyphosate in 11.1% of the total participants. The herbicide was detected more frequently in the neonate population (30%). Multivariable regression models failed to identify significant associations of log-transformed glyphosate with any of the kidney injury biomarkers, controlling for covariates age, sex, and maternal education. While we confirm detectability of glyphosate in children's urine at various ages and stages of life, there is no evidence in this study for renal injury in children exposed to low levels of glyphosate. Further studies of larger sample size are indicated to better understand putative deleterious effects of the herbicide after different levels of exposure.
Product Description
Millipore
MILLIPLEX® Human Kidney Injury Magnetic Bead Panel 1 - Toxicity Multiplex Assay, The analytes available for this multiplex kit are: Calbindin, Collagen IV, Fatty Acid Binding Protein 1 (FABP1), GSTα, IP-10, Kidney Injury Molecule-1 (KIM-1), Renin, TFF-3, TIMP-1.