MilliporeSigma
  • Plasma interleukin-37 is increased and inhibits the production of inflammatory cytokines in peripheral blood mononuclear cells in systemic juvenile idiopathic arthritis patients.
Plasma interleukin-37 is increased and inhibits the production of inflammatory cytokines in peripheral blood mononuclear cells in systemic juvenile idiopathic arthritis patients.
Journal of translational medicine (2018-10-12)
Miao Feng, Min Kang, Feng He, Zonghui Xiao, Zhewei Liu, Hailan Yao, Jianxin Wu
ABSTRACT
Interleukin (IL)-37 has emerged as a novel anti-inflammatory cytokine that play an immunosuppressive role in regulating inflammatory response. This study aimed to measure IL-37 levels in the plasma and peripheral blood mononuclear cells (PBMCs) of patients with systemic juvenile idiopathic arthritis (sJIA), and to establish the correlation between IL-37 levels and disease activity, laboratory parameters and inflammatory cytokines. The mRNA levels of IL-37 in PBMCs and plasma IL-37 concentrations in 46 sJIA patients and 30 age- and sex-matched healthy controls were measured by real-time polymerase chain reaction (RT-PCR) and enzyme-linked immunosorbent assay (ELISA), respectively. The correlations between plasma IL-37 levels and disease activity, laboratory parameters and inflammatory cytokines in sJIA were analyzed by Spearman correlation test. PBMCs from the sJIA patients were stimulated with recombinant human IL-37 (rhIL-37) protein, expressions of IL-1β, IL-6, TNF-α and IL-17 were detected by RT-PCR and ELISA. Plasma levels of IL-37 and relative IL-37 mRNA expression were significantly elevated in sJIA patients, especially in active sJIA patients, when compared with the healthy controls (P < 0.001). Furthermore, patients with active disease showed higher IL-37 mRNAs and plasma protein levels than those with inactive disease as well as healthy controls. Plasma IL-37 levels were correlated with disease activity and inflammatory cytokines (IL-6, TNF-α, IL-17 and GM-CSF) in sJIA patients. The productions of inflammatory cytokines such as IL-6, TNF-α, IL-17 in PBMCs from sJIA patients were obviously decreased after recombinant IL-37 stimulation, whereas the production of IL-1β was not changed. Our results demonstrate that levels of IL-37 were higher in sJIA patients, which were correlated with disease activity and sJIA related inflammatory cytokines. In addition, rhIL-37 down-regulates the expressions of inflammatory cytokines form PBMCs in sJIA patients, suggesting that IL-37 may have the potential role as a natural inhibitor for the pathogenesis and therapy of sJIA.
Product Description
Millipore
MILLIPLEX® Human High Sensitivity T Cell Panel Premixed 13-plex - Immunology Multiplex Assay, Simultaneous analyze low levels of cytokine and chemokine biomarker with the High Sensitivity Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.