MilliporeSigma
Oxidation of the alarmin IL-33 regulates ST2-dependent inflammation.
Nature communications (2015-09-15)
E Suzanne Cohen, Ian C Scott, Jayesh B Majithiya, Laura Rapley, Benjamin P Kemp, Elizabeth England, D Gareth Rees, Catherine L Overed-Sayer, Joanne Woods, Nicholas J Bond, Christel Séguy Veyssier, Kevin J Embrey, Dorothy A Sims, Michael R Snaith, Katherine A Vousden, Martin D Strain, Denice T Y Chan, Sara Carmen, Catherine E Huntington, Liz Flavell, Jianqing Xu, Bojana Popovic, Christopher E Brightling, Tristan J Vaughan, Robin Butler, David C Lowe, Daniel R Higazi, Dominic J Corkill, Richard D May, Matthew A Sleeman, Tomas Mustelin
ABSTRACT
In response to infections and irritants, the respiratory epithelium releases the alarmin interleukin (IL)-33 to elicit a rapid immune response. However, little is known about the regulation of IL-33 following its release. Here we report that the biological activity of IL-33 at its receptor ST2 is rapidly terminated in the extracellular environment by the formation of two disulphide bridges, resulting in an extensive conformational change that disrupts the ST2 binding site. Both reduced (active) and disulphide bonded (inactive) forms of IL-33 can be detected in lung lavage samples from mice challenged with Alternaria extract and in sputum from patients with moderate-severe asthma. We propose that this mechanism for the rapid inactivation of secreted IL-33 constitutes a 'molecular clock' that limits the range and duration of ST2-dependent immunological responses to airway stimuli. Other IL-1 family members are also susceptible to cysteine oxidation changes that could regulate their activity and systemic exposure through a similar mechanism.
Product Description
Millipore
MILLIPLEX® Human TH17 Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple Th17 cytokine and chemokine biomarkers with the Th17 Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.