MilliporeSigma
  • Complex ciprofloxacin resistome revealed by screening a Pseudomonas aeruginosa mutant library for altered susceptibility.
Complex ciprofloxacin resistome revealed by screening a Pseudomonas aeruginosa mutant library for altered susceptibility.
Antimicrobial agents and chemotherapy (2008-10-01)
Elena B M Breidenstein, Bhavjinder K Khaira, Irith Wiegand, Joerg Overhage, Robert E W Hancock
ABSTRACT
Pseudomonas aeruginosa offers substantial therapeutic challenges due to its high intrinsic resistance to many antibiotics and its propensity to develop mutational and/or adaptive resistance. The PA14 comprehensive mutant library was screened for mutants exhibiting either two- to eightfold increased susceptibilities (revealing genes involved in intrinsic resistance) or decreased susceptibilities (mutational resistance) to the fluoroquinolone ciprofloxacin. Thirty-five and 79 mutants with increased and decreased susceptibilities, respectively, were identified, as confirmed by broth dilution.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)