MilliporeSigma
  • G3BP1 controls the senescence-associated secretome and its impact on cancer progression.
G3BP1 controls the senescence-associated secretome and its impact on cancer progression.
Nature communications (2020-10-07)
Amr Omer, Monica Cruz Barrera, Julian L Moran, Xian J Lian, Sergio Di Marco, Christian Beausejour, Imed-Eddine Gallouzi
ABSTRACT
Cellular senescence is a known driver of carcinogenesis and age-related diseases, yet senescence is required for various physiological processes. However, the mechanisms and factors that control the negative effects of senescence while retaining its benefits are still elusive. Here, we show that the rasGAP SH3-binding protein 1 (G3BP1) is required for the activation of the senescent-associated secretory phenotype (SASP). During senescence, G3BP1 achieves this effect by promoting the association of the cyclic GMP-AMP synthase (cGAS) with cytosolic chromatin fragments. In turn, G3BP1, through cGAS, activates the NF-κB and STAT3 pathways, promoting SASP expression and secretion. G3BP1 depletion or pharmacological inhibition impairs the cGAS-pathway preventing the expression of SASP factors without affecting cell commitment to senescence. These SASPless senescent cells impair senescence-mediated growth of cancer cells in vitro and tumor growth in vivo. Our data reveal that G3BP1 is required for SASP expression and that SASP secretion is a primary mediator of senescence-associated tumor growth.
Sigma-Aldrich
MISSION® esiRNA, targeting human G3BP1
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests