MilliporeSigma
  • Characterization of the carbapenem-hydrolyzing oxacillinase oxa-58 in an Acinetobacter genospecies 3 clinical isolate.
Characterization of the carbapenem-hydrolyzing oxacillinase oxa-58 in an Acinetobacter genospecies 3 clinical isolate.
Antimicrobial agents and chemotherapy (2008-05-29)
Sara Marti, Javier Sánchez-Céspedes, M Dolores Blasco, Marc Ruiz, Paula Espinal, Verónica Alba, Felipe Fernández-Cuenca, Alvaro Pascual, Jordi Vila
ABSTRACT
Based on imipenem resistance in an Acinetobacter genospecies 3 clinical isolate, we were able to identify, for the first time in this genomic species, a plasmid-encoded bla(OXA-58) gene that was 100% homologous to the same gene in Acinetobacter baumannii.
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)