MilliporeSigma
  • GABA-B1 Receptor-Null Schwann Cells Exhibit Compromised In Vitro Myelination.
GABA-B1 Receptor-Null Schwann Cells Exhibit Compromised In Vitro Myelination.
Molecular neurobiology (2018-06-28)
Alessandro Faroni, Simona Melfi, Luca Franco Castelnovo, Veronica Bonalume, Deborah Colleoni, Paolo Magni, Marcos J Araúzo-Bravo, Rolland Reinbold, Valerio Magnaghi
ABSTRACT
GABA-B receptors are important for Schwann cell (SC) commitment to a non-myelinating phenotype during development. However, the P0-GABA-B1fl/fl conditional knockout mice, lacking the GABA-B1 receptor specifically in SCs, also presented axon modifications, suggesting SC non-autonomous effects through the neuronal compartment. In this in vitro study, we evaluated whether the specific deletion of the GABA-B1 receptor in SCs may induce autonomous or non-autonomous cross-changes in sensory dorsal root ganglia (DRG) neurons. To this end, we performed an in vitro biomolecular and transcriptomic analysis of SC and DRG neuron primary cultures from P0-GABA-B1fl/fl mice. We found that cells from conditional P0-GABA-B1fl/fl mice exhibited proliferative, migratory and myelinating alterations. Moreover, we found transcriptomic changes in novel molecules that are involved in peripheral neuron-SC interaction.
Sigma-Aldrich
Anti-Neurofilament 160/200 antibody, Mouse monoclonal, ~2 mg/mL, clone RMdO20, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Calcitonin Gene Related Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®