MilliporeSigma
  • Reciprocal Feedback Between miR-181a and E2/ERα in Myometrium Enhances Inflammation Leading to Labor.
Reciprocal Feedback Between miR-181a and E2/ERα in Myometrium Enhances Inflammation Leading to Labor.
The Journal of clinical endocrinology and metabolism (2016-07-28)
Lu Gao, Gang Wang, Wei-Na Liu, Holly Kinser, Hector L Franco, Carole R Mendelson
ABSTRACT
The initiation of term and preterm labor is associated with an up-regulated inflammatory response in myometrium; however, the underlying signaling pathways remain incompletely defined. To define the regulatory mechanisms that mediate the increased myometrial inflammatory response leading to labor, we investigated the roles of microRNAs (miRNA/miR). Human myometrial tissues, isolated smooth muscle cells, and animal models were used to study miR-181a regulation of uterine inflammatory pathways and contractility. Myometrial tissues from 15 term pregnant women undergoing elective cesarean section (not in labor) and 10 term pregnant women undergoing emergency cesarean section (in labor) were used. Expression of the highly conserved microRNA, miR-181a, was significantly decreased in mouse and human myometrium during late gestation. By contrast, the putative miR-181a targets, TNF-α, and estrogen receptor (ER)-α, and the validated target, c-Fos, key factors in the inflammatory response leading to parturition, were coordinately up-regulated. In studies using human myometrial cells, overexpression of miR-181a mimics repressed basal as well as IL-1β-induced TNF-α, C-C motif chemokine ligand 2 and 8 expression, whereas the expression of the antiinflammatory cytokine, IL-10, was increased. Overexpression of miR-181a dramatically inhibited both spontaneous and IL-1β-induced contraction of human myometrial cells. Notably, miR-181a directly targeted ERα and decreased its expression, whereas estradiol-17β reciprocally inhibited expression of mature miR-181a in myometrial cells. Thus, increased estradiol-17β/ERα signaling in myometrium near term inhibits miR-181a, resulting in a further increase in ERα and proinflammatory signaling. This escalating feedback loop provides novel targets and therapeutic strategies for the prevention of preterm labor and its consequences.
Sigma-Aldrich
Anti-RNA polymerase II Antibody, clone CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.
Sigma-Aldrich
ChIPAb+ Estrogen Receptor α - ChIP Validated Antibody and Primer Set, ascites fluid, from mouse