MilliporeSigma
Pick bodies in a family with presenilin-1 Alzheimer's disease.
Annals of neurology (2004-12-29)
Glenda M Halliday, Yun Ju C Song, Gila Lepar, William S Brooks, John B Kwok, Cindy Kersaitis, Gillian Gregory, Claire E Shepherd, Farid Rahimi, Peter R Schofield, Jillian J Kril
ABSTRACT
Presenilin-1 (PS-1) mutations can cause Pick's disease without evidence of Alzheimer's disease (AD). We describe a family with a PS-1 M146L mutation and both Pick bodies and AD. Sarkosyl-insoluble hyperphosphorylated tau showed three bands consistent with AD, although dephosphorylation showed primarily three-repeat isoforms. M146L mutant PS-1 may predispose to both Pick's disease and AD by affecting multiple intracellular pathways involving tau phosphorylation and amyloid metabolism.
Sigma-Aldrich
Monoclonal Anti-τ (Tau) antibody produced in mouse, clone TAU-2, ascites fluid