MilliporeSigma
  • Exogenous application of molybdenum affects the expression of CBF14 and the development of frost tolerance in wheat.
Exogenous application of molybdenum affects the expression of CBF14 and the development of frost tolerance in wheat.
Plant physiology and biochemistry : PPB (2012-12-19)
Mohammed Al-Issawi, Hail Z Rihan, Wondwossen Abate Woldie, Stephen Burchett, Michael P Fuller
ABSTRACT
Wheat is able to cold acclimate in response to low temperatures and thereby increase its frost tolerance and the extent of this acclimation is greater in winter genotypes compared to spring genotypes. Such up-regulation of frost tolerance is controlled by Cbf transcription factors. Molybdenum (Mo) application has been shown to enhance frost tolerance of wheat and this study aimed to investigate the effect of Mo on the development of frost tolerance in winter and spring wheat. Results showed that Mo treatment increased the expression of Cbf14 in wheat under non-acclimating condition but did not alter frost tolerance. However, when Mo was applied in conjunction with exposure of plants to low temperature, Mo increased the expression of Cbf14 and enhanced frost tolerance in both spring and winter genotypes but the effect was more pronounced in the winter genotype. It was concluded that the application of Mo could be useful in situations where enhanced frost resistance is required. Further studies are proposed to elucidate the effect of exogenous of applications of Mo on frost resistance in spring and winter wheat at different growth stages.
Sigma-Aldrich
Spectrum Plant Total RNA Kit, sufficient for 50 purifications