MilliporeSigma
  • PRIMA-1(MET) induces nucleolar accumulation of mutant p53 and PML nuclear body-associated proteins.
PRIMA-1(MET) induces nucleolar accumulation of mutant p53 and PML nuclear body-associated proteins.
Oncogene (2006-08-16)
N Rökaeus, G Klein, K G Wiman, L Szekely, K Mattsson
ABSTRACT
We have previously identified PRIMA-1, a low molecular weight compound that restores the transcriptional transactivation function to mutant p53 and induction of apoptosis. To explore the molecular mechanism for PRIMA-1-induced mutant p53-dependent apoptosis, we examined the intracellular distribution of mutant p53 upon treatment with PRIMA-1(MET) by immunofluorescence staining. We found that PRIMA-1(MET) induced nucleolar translocation of mutant p53 and the promyelocytic leukemia (PML) nuclear body-associated proteins PML, CBP and Hsp70. Levels of Hsp70 were significantly enhanced by PRIMA-1(MET) treatment. PRIMA-Dead, a compound structurally related to PRIMA-1 but unable to induce mutant p53-dependent apoptosis, failed to induce nucleolar translocation of mutant p53. Our results suggest that redistribution of mutant p53 to nucleoli plays a role in PRIMA-1-induced apoptosis.
Sigma-Aldrich
CaspaTag Pan-Caspase In Situ Assay Kit, Fluorescein, The In Situ FLICA Pan-Caspase Detection Kit for flow cytometry is a fluorescent-based assay for detection of active caspases in cells undergoing apoptosis.