MilliporeSigma
  • SF3B1 mutant MDS-initiating cells may arise from the haematopoietic stem cell compartment.
SF3B1 mutant MDS-initiating cells may arise from the haematopoietic stem cell compartment.
Nature communications (2015-12-09)
Syed A Mian, Kevin Rouault-Pierre, Alexander E Smith, Thomas Seidl, Irene Pizzitola, Aytug Kizilors, Austin G Kulasekararaj, Dominique Bonnet, Ghulam J Mufti
ABSTRACT
Despite the recent evidence of the existence of myelodysplastic syndrome (MDS) stem cells in 5q-MDS patients, it is unclear whether haematopoietic stem cells (HSCs) could also be the initiating cells in other MDS subgroups. Here we demonstrate that SF3B1 mutation(s) in our cohort of MDS patients with ring sideroblasts can arise from CD34(+)CD38(-)CD45RA(-)CD90(+)CD49f(+) HSCs and is an initiating event in disease pathogenesis. Xenotransplantation of SF3B1 mutant HSCs leads to persistent long-term engraftment restricted to myeloid lineage. Moreover, genetically diverse evolving subclones of mutant SF3B1 exist in mice, indicating a branching multi-clonal as well as ancestral evolutionary paradigm. Subclonal evolution in mice is also seen in the clinical evolution in patients. Sequential sample analysis shows clonal evolution and selection of the malignant driving clone leading to AML transformation. In conclusion, our data show SF3B1 mutations can propagate from HSCs to myeloid progeny, therefore providing a therapeutic target.
Sigma-Aldrich
SeqPlex DNA Amplification Kit, For use with high throughput sequencing technologies, Whole Genome Amplification kit designed to facilitate Next Gen Sequencing.
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 reactions
Please contact Customer Service for Availability
$396.00
50 reactions
Please contact Customer Service for Availability
$1,510.00
500 reactions
Please contact Customer Service for Availability
$10,960.00
Sigma-Aldrich
GenomePlex® Single Cell Whole Genome Amplification Kit, Amplify genome of a single cell
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 reactions
Please contact Customer Service for Availability
$347.00
50 reactions
Please contact Customer Service for Availability
$1,410.00
500 reactions
Please contact Customer Service for Availability
$9,310.00