MilliporeSigma
  • Xanthine oxidase mediates hypoxia-inducible factor-2α degradation by intermittent hypoxia.
Xanthine oxidase mediates hypoxia-inducible factor-2α degradation by intermittent hypoxia.
PloS one (2013-10-15)
Jayasri Nanduri, Damodara Reddy Vaddi, Shakil A Khan, Ning Wang, Vladislav Makerenko, Nanduri R Prabhakar
ABSTRACT
Sleep-disordered breathing with recurrent apnea produces chronic intermittent hypoxia (IH). We previously reported that IH leads to down-regulation of HIF-2α protein via a calpain-dependent signaling pathway resulting in oxidative stress. In the present study, we delineated the signaling pathways associated with calpain-dependent HIF-2α degradation in cell cultures and rats subjected to chronic IH. Reactive oxygen species (ROS) scavengers prevented HIF-2α degradation by IH and ROS mimetic decreased HIF-2α protein levels in rat pheochromocytoma PC12 cell cultures, suggesting that ROS mediate IH-induced HIF-2α degradation. IH activated xanthine oxidase (XO) by increased proteolytic conversion of xanthine dehydrogenase to XO. ROS generated by XO activated calpains, which contributed to HIF-2α degradation by IH. Calpain-induced HIF-2α degradation involves C-terminus but not the N-terminus of the HIF-2α protein. Pharmacological blockade as well as genetic knock down of XO prevented IH induced calpain activation and HIF-2α degradation in PC12 cells. Systemic administration of allopurinol to rats prevented IH-induced hypertension, oxidative stress and XO activation in adrenal medulla. These results demonstrate that ROS generated by XO activation mediates IH-induced HIF-2α degradation via activation of calpains.
Sigma-Aldrich
InnoZyme Calpain 1/2 Activity Assay Kit