MilliporeSigma
  • Chronic morphine up-regulates G alpha12 and cytoskeletal proteins in Chinese hamster ovary cells expressing the cloned mu opioid receptor.
Chronic morphine up-regulates G alpha12 and cytoskeletal proteins in Chinese hamster ovary cells expressing the cloned mu opioid receptor.
The Journal of pharmacology and experimental therapeutics (2005-07-01)
Heng Xu, Xiaoying Wang, Darin Zimmerman, Emily S Boja, Jiabei Wang, Edward J Bilsky, Richard B Rothman
ABSTRACT
A growing body of literature indicates that chronic morphine exposure alters the expression and function of cytoskeletal proteins in addition to the well established interactions between mu opioid receptors and G proteins. In the present study, we hypothesized that chronic morphine alters the expression and functional effects of G alpha12, a G protein that regulates downstream cytoskeletal proteins via its control of RhoA. Our results showed that chronic morphine treatment decreased the expression of G alpha i2 (64%) and G alpha i3 (60%), had no effect of G alpha o, and increased G alpha12 (66%) expression in Chinese hamster ovary (CHO) cells expressing the cloned human mu opioid receptors (hMOR-CHO cells) but not in cells expressing a mutant mu opioid receptor that do not develop morphine tolerance and dependence (T394A-CHO cells). Morphine treatment had no significant effect on PAR-1 thrombin receptor-activated G protein activity, as measured by thrombin-stimulated guanosine 5'-O-(3-[35S]thio)triphosphate binding. Chronic morphine treatment significantly enhanced thrombin-stimulated RhoA activity and thrombin-stimulated expression of alpha-actinin, a cytoskeletal anchoring protein, in hMOR-CHO cells. Proteomic analysis of two-dimensional gel spots prepared from hMOR-CHO cells showed that morphine treatment affected the expression of a number of proteins associated with morphological changes. Up-regulation of G alpha12 and alpha-actinin by chronic morphine was also observed in mouse brain. Viewed collectively, these findings indicate, for the first time, that chronic morphine enhances the G alpha12-associated signaling system, which is involved in regulating cellular morphology and growth, supporting other findings that chronic morphine may alter cellular morphology, in addition to cellular function.
Sigma-Aldrich
Anti-Opioid μ Receptor (384-398) Rabbit pAb, lyophilized, Calbiochem®