MilliporeSigma
  • Differences in Alimentary Glucose Absorption and Intestinal Disposal of Blood Glucose After Roux-en-Y Gastric Bypass vs Sleeve Gastrectomy.
Differences in Alimentary Glucose Absorption and Intestinal Disposal of Blood Glucose After Roux-en-Y Gastric Bypass vs Sleeve Gastrectomy.
Gastroenterology (2015-10-21)
Jean-Baptiste Cavin, Anne Couvelard, Rachida Lebtahi, Robert Ducroc, Konstantinos Arapis, Eglantine Voitellier, Françoise Cluzeaud, Laura Gillard, Muriel Hourseau, Nidaa Mikail, Lara Ribeiro-Parenti, Nathalie Kapel, Jean-Pierre Marmuse, André Bado, Maude Le Gall
ABSTRACT
Bariatric procedures, such as Roux-en-Y gastric bypass (RYGB) or vertical sleeve gastrectomy (VSG), are the most effective approaches to resolve type 2 diabetes in obese individuals. Alimentary glucose absorption and intestinal disposal of blood glucose have not been directly compared between individuals or animals that underwent RYGB vs VSG. We evaluated in rats and humans how the gut epithelium adapts after surgery and the consequences on alimentary glucose absorption and intestinal disposal of blood glucose. Obese male rats underwent RYGB, VSG, or sham (control) operations. We collected intestine segments from all rats; we performed histologic analyses and measured levels of messenger RNAs encoding the sugar transporters SGLT1, GLUT1, GLUT2, GLUT3, GLUT4, and GLUT5. Glucose transport and consumption were assayed using ex vivo jejunal loops. Histologic analyses were also performed on Roux limb sections from patients who underwent RYGB 1-5 years after surgery. Roux limb glucose consumption was assayed after surgery by positron emission and computed tomography imaging. In rats and humans that underwent RYGB, the Roux limb became hyperplasic, with an increased number of incretin-producing cells compared with the corresponding jejunal segment of controls. Furthermore, expression of sugar transporters and hypoxia-related genes increased and the nonintestinal glucose transporter GLUT1 appeared at the basolateral membrane of enterocytes. Ingested and circulating glucose was trapped within the intestinal epithelial cells of rats and humans that underwent RYGB. By contrast, there was no hyperplasia of the intestine after VSG, but the intestinal absorption of alimentary glucose was reduced and density of endocrine cells secreting glucagon-like peptide-1 increased. The intestine adapts differently to RYGB vs VSG. RYGB increases intestinal glucose disposal and VSG delays glucose absorption; both contribute to observed improvements in glycemia.