MilliporeSigma
  • CK2 kinase-mediated PHF8 phosphorylation controls TopBP1 stability to regulate DNA replication.
CK2 kinase-mediated PHF8 phosphorylation controls TopBP1 stability to regulate DNA replication.
Nucleic acids research (2020-10-04)
Haihua Feng, Jingchen Lu, Xiaotian Song, Angkana Thongkum, Fan Zhang, Lihong Lou, Ofer Reizes, Alexandru Almasan, Zihua Gong
ABSTRACT
ATR functions as a master regulator of the DNA-damage response. ATR activation requires the ATR activator, topoisomerase IIβ-binding protein 1 (TopBP1). However, the underlying mechanism of TopBP1 regulation and how its regulation affects DNA replication remain unknown. Here, we report a specific interaction between TopBP1 and the histone demethylase PHF8. The TopBP1/PHF8 interaction is mediated by the BRCT 7+8 domain of TopBP1 and phosphorylation of PHF8 at Ser854. This interaction is cell-cycle regulated and phosphorylation-dependent. PHF8 is phosphorylated by CK2, which regulates binding of PHF8 to TopBP1. Importantly, PHF8 regulates TopBP1 protein level by preventing its ubiquitination and degradation mediated by the E3 ligase UBR5. Interestingly, PHF8pS854 is likely to contribute to regulation of TopBP1 stability and DNA replication checkpoint. Further, both TopBP1 and PHF8 are required for efficient replication fork restart. Together, these data identify PHF8 as a TopBP1-binding protein and provide mechanistic insight into how PHF8 regulates TopBP1 stability to maintain DNA replication.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Histone H3 Antibody, CT, pan, clone A3S, rabbit monoclonal, culture supernatant, clone A3S, Upstate®
Sigma-Aldrich
Anti-Maltose Binding Protein Antibody, serum, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid