MilliporeSigma
  • What is the effect of a Mediterranean compared with a Fast Food meal on the exercise induced adipokine changes? A randomized cross-over clinical trial.
What is the effect of a Mediterranean compared with a Fast Food meal on the exercise induced adipokine changes? A randomized cross-over clinical trial.
PloS one (2019-04-19)
Diana Silva, Rita Moreira, Marília Beltrão, Oksana Sokhatska, Tiago Montanha, Andreia Pizarro, Vanessa Garcia-Larsen, Rodrigo Villegas, Luís Delgado, Pedro Moreira, Joana Carvalho, André Moreira
ABSTRACT
Adipose tissue-derived adipokines are pro-inflammatory cytokines involved in metabolic-related diseases and can be influenced by diet and exercise. We aimed to compare the effect of a Mediterranean (MdM) compared with Fast Food (FFM) meal on the exercise induced adipokines changes. In a double blinded cross over trial, 46 participants were randomly assigned to one of two standardized iso-energy pre-exercise meals: FFM or MdM-type. Three hours after each meal, participants completed a treadmill exercise test (EC). Serum adiponectin, resistin, PAI-1, lipocalin-2/NGAL and adipsin were determined by Luminex magnetic bead immunoassay. Wilcoxon signed rank test compared changes before/after meal and before/after EC and a linear mixed model evaluated the effect of meals on the adipokine response to exercise, adjusted for confounders. Thirty-nine participants (mean age of 25, with a standard deviation of 5 years) completed the trial (56% females). For both interventions, a significant reduction of adipsin after each meal and a significant increase of lipocalin, PAI-1, adipsin and resistin, after exercise was observed. When exercise was preceded by a MdM meal a higher increase in adipsin levels was seen. Acute exercise induced an increase of circulatory levels of adipsin, resistin, lipocalin and PAI-1, but not adiponectin. A pre-exercise Mediterranean meal potentiated the increase of adipsin after the exercise test, which possibly relates to the immune regulatory role of adipsin. These changes suggest a cross-talk between the immune and metabolic immediate response to exercise and its modulation by the pre-exercise diet composition.
Product Description
Millipore
MILLIPLEX® Human Adipokine Magnetic Bead Panel 1 - Endocrine Multiplex Assay, The Human Adipokine Panel 1, using the Luminex xMAP technology, enables the simultaneous analysis of five biomarkers in human serum or plasma samples.