MilliporeSigma
  • Mouse neutrophils express the decoy type 2 interleukin-1 receptor (IL-1R2) constitutively and in acute inflammatory conditions.
Mouse neutrophils express the decoy type 2 interleukin-1 receptor (IL-1R2) constitutively and in acute inflammatory conditions.
Journal of leukocyte biology (2013-07-03)
Praxedis Martin, Gaby Palmer, Solenne Vigne, Céline Lamacchia, Emiliana Rodriguez, Dominique Talabot-Ayer, Stefan Rose-John, Athena Chalaris, Cem Gabay
ABSTRACT
The proinflammatory activities of IL-1 are tightly controlled at different levels. IL-1R2 acts as a decoy receptor and has been shown to regulate the biological effects of IL-1 in vitro and in vivo. However, little is known about its natural expression in the mouse in physiologic and pathologic conditions. In this study, we examined IL-1R2 mRNA and protein expression in isolated cells and tissues in response to different stimulatory conditions. Data obtained using ex vivo CD11b(+)Ly6G(+) peripheral blood cells and in vitro-differentiated CD11b(+)Ly6G(+) BMG indicated that neutrophils are the major source of constitutively expressed IL-1R2 in the mouse. The expression of IL-1R2 on BMG and ex vivo Ly6G(+) peripheral blood cells was highly up-regulated by HC. IL-1R2 pull-down experiments showed that mouse rIL-1β binds to BMG IL-1R2, whereas binding of IL-1Ra could not be detected. Furthermore, LPS treatment induced shedding of IL-1R2 from the neutrophil membrane in vitro and in vivo, executed mainly by ADAM17. Finally, in in vivo models of inflammation, including thioglycolate-induced acute peritonitis and acute lung injury, infiltrating Ly6G(+) neutrophils, expressed IL-1R2. Our data show that in the mouse, neutrophils mainly express the decoy receptor IL-1R2 under naïve and inflammatory conditions. These data suggest that neutrophils may contribute to the resolution of acute inflammation.