MilliporeSigma
Application and microbial preparation of D-valine.
World journal of microbiology & biotechnology (2016-08-28)
Ming Chen, Chao Shi, Jing Zhao, Ziqing Gao, Chunzhi Zhang
ABSTRACT
D-Valine is an important organic chiral source and has extensive industrial application, which is used as intermediate for the synthesis of agricultural pesticides, semi-synthetic veterinary antibiotics and pharmaceutical drugs. Its derivatives have shown great activity in clinical use, such as penicillamine for the treatment of immune-deficiency diseases, and actinomycin D for antitumor therapy. Fluvalinate, a pyrethroid pesticide made from D-valine, is a broad-spectrum insecticide with low mammalian toxicity. Valnemulin, a semi-synthetic pleuromutilin derivative synthesized from D-valine, is an antibiotic for animals. Moreover, D-valine is also used in cell culture for selectively inhibiting fibroblasts proliferation. Due to its widespread application, D-valine is gaining more and more attention and some approaches for D-valine preparation have been investigated. In comparison with other approaches, microbial preparation of D-valine is more competitive and promising because of its high stereo selectivity, mild reaction conditions and environmental friendly process. So far, microbial preparation of D-valine can be mainly classified into three categories: microbial asymmetric degradation of DL-valine, microbial stereoselective hydrolysis of N-acyl-DL-valine by D-aminoacylase, and microbial specific hydrolysis of DL-5-isopropylhydantoin by D-hydantoinase coupled with D-carbamoylase. In this paper, the industrial application of D-valine and its microbial preparation are reviewed.
Sigma-Aldrich
D-Valine, suitable for cell culture, BioReagent