MilliporeSigma
  • Structure-activity relationship of flavonoids as influenza virus neuraminidase inhibitors and their in vitro anti-viral activities.
Structure-activity relationship of flavonoids as influenza virus neuraminidase inhibitors and their in vitro anti-viral activities.
Bioorganic & medicinal chemistry (2008-07-22)
Ai-Lin Liu, Hai-Di Wang, Simon MingYuen Lee, Yi-Tao Wang, Guan-Hua Du
ABSTRACT
Flavonoids are polyphenolic compounds that widely exist in plant kingdom, and the structure-activity relationship (SAR) of 25 flavonoids was studied on neuraminidase (NA) activity of influenza virus. Three typical influenza virus strains A/PR/8/34 (H1N1), A/Jinan/15/90 (H3N2), and B/Jiangshu/10/2003 were used as the source of NAs, the average of IC(50)s of these compounds on these NAs was used in the SAR analysis. The order of potency for NA inhibition was as follows: aurones>flavon(ol)es>isoflavones>flavanon(ol)es and flavan(ol)es. The SAR analysis of flavonoids on influenza virus NAs revealed that for good inhibitory effect, the 4'-OH, 7-OH, C4O, and C2C3 functionalities were essential, and the presence of a glycosylation group greatly reduced NA inhibition. The in vitro anti-viral activities of eight flavonoids were evaluated using a cytopathic effect (CPE) reduction method, the assay results confirmed the SAR as influenza virus neuraminidase inhibitors. The findings of this study provide important information for the exploitation and utilization of flavonoids as NA inhibitors for influenza treatment.
Sigma-Aldrich
Apigenin, ≥95.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
Luteolin, ≥98% (TLC), powder
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Chrysin, ≥96.5%