MilliporeSigma
  • Association of Urinary Dickkopf-3 with Residual Renal Function Decline in Patients Undergoing Peritoneal Dialysis.
Association of Urinary Dickkopf-3 with Residual Renal Function Decline in Patients Undergoing Peritoneal Dialysis.
Medicina (Kaunas, Lithuania) (2021-07-03)
Kenta Torigoe, Kumiko Muta, Kiyokazu Tsuji, Ayuko Yamashita, Miki Torigoe, Shinichi Abe, Yuki Ota, Hiroshi Mukae, Tomoya Nishino
ABSTRACT
Background and Objectives: Urinary levels of dickkopf-3 (DKK-3) are associated with poor renal survival in patients with non-dialytic chronic kidney disease. However, it remains unknown whether urinary DKK-3 levels can predict residual renal function (RRF) decline in patients undergoing peritoneal dialysis (PD). Therefore, we investigated the correlation between urinary levels of DKK-3 and the subsequent rate of RRF decline in PD patients. Materials and Methods: This study included 36 PD patients who underwent multiple peritoneal equivalent tests during 2011-2021. The relationship between baseline clinical characteristics and the subsequent annual rate of Kt/V decline was investigated. Results: The annual rate of renal Kt/V decline was 0.29 (range: 0.05-0.48), which correlated with renal Kt/V (r = 0.55, p = 0.0005) and 24 h urinary DKK-3 excretion (r = 0.61, p < 0.0001). Similarly, 24 h urinary DKK-3 excretion (β = 0.44, p = 0.0015) and renal Kt/V (β = 0.38, p = 0.0059) were independently associated with the annual rate of renal Kt/V decline in multivariate analyses. Conclusions: Urinary DKK-3 assessment may help identify PD patients at a high risk of RRF decline.
Sigma-Aldrich
Human Dkk-3 ELISA Kit, for serum, plasma, cell culture supernatants and urine