MilliporeSigma
  • The functions and properties of cullin-5, a potential therapeutic target for cancers.
The functions and properties of cullin-5, a potential therapeutic target for cancers.
American journal of translational research (2020-03-21)
Feng Gao, Yimin Fan, Bolun Zhou, Weihua Guo, Xingjun Jiang, Jing Shi, Caiping Ren
ABSTRACT
Cullin-5 (CUL5), a scaffold protein in active cullin-RING ubiquitin ligase (CRL) complexes, is a member of the cullin family of proteins. The CUL5-type ubiquitin ligase can target multiple proteins involved in ubiquitination and proteasome degradation. CUL5 plays positive roles in regulating cell growth, proliferation and physiological and other processes in the human body. It has been found that the expression of CUL5 is significantly downregulated in various cancer cells, which affects the course of the cancers. Here, we reviewed the current data on the expression and role of CUL5 in both normal and cancer cells, its possible mechanisms, and its potential as a therapeutic target for cancers.