MilliporeSigma
  • DNA interference: a simple and efficient gene-silencing system for high-throughput functional analysis in the fern adiantum.
DNA interference: a simple and efficient gene-silencing system for high-throughput functional analysis in the fern adiantum.
Plant & cell physiology (2004-12-03)
Hiroko Kawai-Toyooka, Chihiro Kuramoto, Kyoko Orui, Kentaro Motoyama, Kazuhiro Kikuchi, Takeshi Kanegae, Masamitsu Wada
ABSTRACT
RNA interference (RNAi) has become a powerful tool for determining gene function and is used in a wide variety of organisms. Since it is necessary to generate double-stranded RNA (dsRNA) as an inducer for RNAi, preparation of RNAi-inducing constructs is somewhat cumbersome and time consuming, especially for the thousands of genes used in a genome-wide analysis. To overcome these problems, we have developed a more convenient gene-silencing method in the fern Adiantum using double-stranded DNA (dsDNA) as a model system for functional analysis in plants. Delivery of dsDNA fragments homologous to an endogenous gene into gametophytic cells can induce sequence-specific gene silencing. As it only requires dsDNA fragments homologous to a target gene, PCR-amplified fragments are enough to trigger gene silencing. Maximum gene silencing efficiencies of >90% have been achieved for transformed plants. In addition, simultaneous transfer of dsDNA fragments corresponding to multiple genes still has a silencing effect for individual genes. We term this approach 'DNA interference'.
Sigma-Aldrich
White′s Basal Salt Mixture, powder, suitable for plant cell culture