MilliporeSigma
  • TBX18 transcription factor overexpression in human-induced pluripotent stem cells increases their differentiation into pacemaker-like cells.
TBX18 transcription factor overexpression in human-induced pluripotent stem cells increases their differentiation into pacemaker-like cells.
Journal of cellular physiology (2018-08-06)
Armita M Gorabi, Saeideh Hajighasemi, Hossein A Tafti, Amir Atashi, Masoud Soleimani, Nasser Aghdami, Ali K Saeid, Vahid Khori, Yunes Panahi, Amirhossein Sahebkar
ABSTRACT
The discovery of gene- and cell-based strategies has opened a new area to investigate novel approaches for the treatment of many conditions caused by cardiac cell failure. The TBX18 (T-box 18) transcription factor is considered as a prominent factor in the sinoatrial node (SAN) formation during the embryonic development. In this in vitro study, the effect of TBX18 gene expression on human-induced pluripotent-stem-cell-derived cardiomyocytes (hiPS-CMs) to induce pacemaker-like cells was examined. The human-dermal-fibroblast-derived iPSCs were transfected using chemical, physical, and Lentiviral methods of TBX18 gene delivery during differentiation into cardiomyocytes (CMs). After the differentiation process through small-molecule-based temporal modulation of the Wnt signaling pathway, the hiPSC-CMs were analyzed using the real-time polymerase chain reaction, immunocytochemistry, immunofluorescence, whole-cell patch-clamp recording, and western blotting to investigate the accuracy of differentiation and identify the effect exerted by TBX18. The hiPS-CMs showed spontaneous beating and expressed specific markers of cardiac cells. The lentiviral-mediated TBX18 delivery was the most efficient method for transfection. The results showed the increment in Connexin 43 expression among untransfected hiPS-CMs, whereas this protein was significantly downregulated followed by TBX18 overexpression. TBX18-hiPSCMs were detected with pacemaker cell features. It was demonstrated that the TBX18 gene is able to conduct hiPSCs to differentiate into pacemaker-like cells. The TBX18 gene delivery seems to have the potential for the development of biological pacemakers; however, more investigations are still needed to assess its usefulness to fix arrhythmic conditions with SAN failure basis.
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal Anti-α-Actinin (Sarcomeric) antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit