MilliporeSigma
  • Human type I pituitary adenylate cyclase activating polypeptide receptor (ADCYAP1R): localization to chromosome band 7p14 and integration into the cytogenetic, physical, and genetic map of chromosome 7.
Human type I pituitary adenylate cyclase activating polypeptide receptor (ADCYAP1R): localization to chromosome band 7p14 and integration into the cytogenetic, physical, and genetic map of chromosome 7.
Genomics (1994-10-01)
M Stoffel, R Espinosa, J B Trabb, M M Le Beau, G I Bell
ABSTRACT
The gene encoding the human type I pituitary adenylate cyclase activating polypeptide receptor (ADCYAP1R1) was mapped to chromosome 7 by PCR analysis of genomic DNA from a human/rodent somatic cell hybrid mapping panel. This assignment was confirmed and the gene localized to chromosome band 7p14 by fluorescence in situ hybridization. A yeast artificial chromosome containing ADCYAP1R1 was identified in the CEPH "B" Mega-YAC library. This YAC includes two highly polymorphic dinucleotide repeat sequences that will facilitate genetic studies of the contribution of ADCYAP1R1 in disease states of the central nervous and neuroendocrine systems.