MilliporeSigma
  • Overexpression of beta 1-adrenoceptors in adult rat ventricular myocytes enhances CGP 12177A cardiostimulation: implications for 'putative' beta 4-adrenoceptor pharmacology.
Overexpression of beta 1-adrenoceptors in adult rat ventricular myocytes enhances CGP 12177A cardiostimulation: implications for 'putative' beta 4-adrenoceptor pharmacology.
British journal of pharmacology (2004-02-06)
Clive J Lewis, Haibin Gong, Morris J Brown, Sian E Harding
ABSTRACT
1. CGP 12177A mediates cardiostimulation by activation of the 'putative' beta(4)-adrenoceptor; however, it has recently been reported that disruption of the beta(1)-adrenoceptor gene abolishes this effect. We have adenovirally overexpressed beta(1)-adrenoceptors in isolated, cultured adult rat ventricular cardiomyocytes and observed the inotropic potency of isoprenaline and CGP 12177A (in the presence of 1 microm propranolol). 2. Isoprenaline was a full inotropic agonist at rat ventricular myocytes (pD(2) 7.69+/-0.12). CGP 12177A was a nonconventional partial agonist (pD(2) 6.34+/-0.09), increasing inotropy and lusitropy, with an intrinsic activity of 0.34 and antagonised by bupranolol. 3. beta(1)-adrenoceptor overexpression enhanced the inotropic potency of isoprenaline by 11.7-fold (pD(2) 8.76+/-0.14) and CGP 12177A by 5.9-fold (7.11+/-0.10), respectively. Green fluorescent protein (GFP) overexpression did not alter the potency of isoprenaline or CGP 12177A (pD(2) 7.41+/-0.24 and pD(2) 6.60+/-0.50, respectively). 4. The cardiostimulant effects of CGP 12177A were enhanced by IBMX (phosphodiesterase inhibitor) and decreased by Rp-cAMPS (cAMP antagonist). CGP 12177A also increased cAMP levels. CGP 12177A but not isoprenaline initiated arrhythmias at lower concentrations following beta(1)-adrenoceptor overexpression. 5. (125)I-Cyanopindolol saturation binding in Adv.beta(1) myocytes demonstrated approximately 18-fold increase in beta(1)-adrenoceptors. (3)H-CGP 12177A saturation binding, in the presence of propranolol, increased approximately 5-fold following overexpression of beta(1)-adrenoceptors. 6. This study demonstrates enhanced cardiostimulation by CGP 12177A (in the presence of propranolol) in rat ventricular myocytes overexpressing beta(1)-adrenoceptors, mediated by a Gs/cAMP signalling pathway. 'Putative' beta(4)-adrenoceptor pharmacology appears to be mediated by activation of a novel affinity state of the beta(1)-adrenoceptor.
Sigma-Aldrich
(±)-CGP-12177A, ≥98% (HPLC), solid