MilliporeSigma
  • AVE 0991, a nonpeptide mimic of the effects of angiotensin-(1-7) on the endothelium.
AVE 0991, a nonpeptide mimic of the effects of angiotensin-(1-7) on the endothelium.
Hypertension (Dallas, Tex. : 1979) (2002-12-07)
Gabriele Wiemer, Lawrence W Dobrucki, Febee R Louka, Tadeusz Malinski, Holger Heitsch
ABSTRACT
Recently, we demonstrated that the heptapeptide angiotensin-(1-7) (Ang-[1-7]) exhibits a favorable kinetic of nitric oxide (NO) release accompanied by extremely low superoxide (O2-) production. In this report we describe AVE 0991, a novel nonpeptide compound that evoked effects similar to Ang-(1-7) on the endothelium. AVE 0991 and unlabeled Ang-(1-7) competed for high-affinity binding of [125I]-Ang-(1-7) to bovine aortic endothelial cell membranes with IC50 values of 21+/-35 and 220+/-280 nmol/L, respectively. Stimulated NO and O2- release from bovine aortic endothelial cells was directly and simultaneously measured on the cell surface by selective electrochemical nanosensors. Peak concentrations of NO and O2- release by AVE 0991 and Ang-(1-7) (both 10 micromol/L) were not significantly different (NO: 295+/-20 and 270+/-25 nmol/L; O2-: 18+/-2 and 20+/-4 nmol/L). However, the released amount of bioactive NO was approximately 5 times higher for AVE 0991 in comparison to Ang-(1-7). The selective Ang-(1-7) antagonist [D-Ala(7)]-Ang-(1-7) inhibited the AVE 0991-induced NO and O2- production by approximately 50%. A similar inhibition level was observed for the Ang II AT1 receptor antagonist EXP 3174. In contrast, the Ang II AT2 receptor antagonist PD 123,177 inhibited the AVE 0991-stimulated NO production by approximately 90% but without any inhibitory effect on O2- production. Both NO and O2- production were inhibited by NO synthase inhibition ( approximately 70%) and by bradykinin B2 receptor blockade (approximately 80%). AVE 0991 efficiently mimics the effects of Ang-(1-7) on the endothelium, most probably through stimulation of a specific, endothelial Ang-(1-7)-sensitive binding site causing kinin-mediated activation of endothelial NO synthase.
Sigma-Aldrich
AVE 0991, ≥98% (HPLC)