MilliporeSigma
  • Co-existence of alpha-glucosidase-inhibitory and liver X receptor-regulatory activities and their separation by structural development.
Co-existence of alpha-glucosidase-inhibitory and liver X receptor-regulatory activities and their separation by structural development.
Bioorganic & medicinal chemistry (2008-03-18)
Kosuke Dodo, Atsushi Aoyama, Tomomi Noguchi-Yachide, Makoto Makishima, Hiroyuki Miyachi, Yuichi Hashimoto
ABSTRACT
Liver X receptors (LXR), which were originally reported as oxysterol-activated nuclear receptors, were recently found to recognize glucose as a physiological ligand. On this basis, we have already developed novel LXR antagonists based upon alpha-glucosidase inhibitors derived from thalidomide. Here, to clarify the relationship between alpha-glucosidase inhibition and LXR modulation, we investigate the alpha-glucosidase-inhibitory activity of typical LXR ligands and the LXR-modulating activity of typical alpha-glucosidase inhibitors. Although there were some exceptions, co-existence of LXR-regulatory and alpha-glucosidase-inhibitory activities seemed to be rather general among the examined compounds. The LXR ligands were found to be non-competitive alpha-glucosidase inhibitors, suggesting that it might be possible to separate the two activities. To test this idea, we focused on riccardin C, a naturally occurring LXR ligand, which we found here to be a potent alpha-glucosidase inhibitor as well. Structural development of riccardin C afforded novel LXR antagonists lacking alpha-glucosidase-inhibitory activity, 19c and 19f, and a LXRalpha-selective antagonist, 22.
Sigma-Aldrich
22(R)-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder