MilliporeSigma
  • A combination of cis-2-decenoic acid and chlorhexidine removes dental plaque.
A combination of cis-2-decenoic acid and chlorhexidine removes dental plaque.
Archives of oral biology (2015-09-10)
Azadeh Rahmani-Badi, Shayesteh Sepehr, Hamta Babaie-Naiej
ABSTRACT
To investigate the ability of cis-2-decenoic acid (C2DA) to induce dispersion in single-species biofilms formed by Streptococcus mutans and Candida albicans, as well as to remove their bacterial-fungal dual-species biofilms when combined with low concentrations of chlorhexidine (CHX). For biofilm dispersal bioassays, single-species biofilms of S. mutans and C. albicans were grown on the inside surface of petri dishes, using a semi-batch culture method in which the medium was replaced every 24h for 5 days. Biofilms were then treated with very low concentrations of C2DA (100 and 310 nM) for 1h to release cells into the bulk liquid and to evaluate dispersed cell number by measuring the optical density (OD). To assess the ability of C2DA combined CHX treatments to remove tested microorganisms' dual-species biofilms, they were grown on saliva-coated hydroxyapatite (sHA) discs for 48 h and then were treated with three different concentrations of CHX (0.08%, 0.06% and 0.04%) alone or in combination with indicated concentrations of C2DA for 1 min twice daily for 3 subsequent days. Biofilms were then either subjected to the field emission scanning electron microscopy (FESEM) analysis or harvested and colony forming units (CFUs) were counted after plating on agar. Treatment of pre-established biofilms with 310 nM C2DA caused an approximately two-fold increase in the number of planktonic cells in both cultures. A combination of 310 nM C2DA and 0.04% CHX resulted in significant removal (p-value <0.05) of dual-species biofilms from sHA discs surface. Anti-biofilm characteristic of C2DA boosts the action of CHX even at low concentrations.
Sigma-Aldrich
cis-2-Decenoic acid, ≥95.0% (HPLC)