MilliporeSigma
  • Examining the Presence of Cronobacter spp. in Ready-to-eat Edible Insects.
Examining the Presence of Cronobacter spp. in Ready-to-eat Edible Insects.
Food safety (Tokyo, Japan) (2020-01-31)
Jake P Greenhalgh, Daniel Amund
ABSTRACT
Edible insects present a potential solution to increasing global food insecurity. However, there is limited research on the microbial hazards they may pose. These include opportunistic pathogens like Cronobacter spp. (formerly Enterobacter sakazakii). In this study, nine types of ready-to-eat edible insect products purchased in the UK were examined for their microbial load (total aerobic count, total Enterobacteriaceae count), and screened for the presence of Cronobacter sakazakii (C. sakazakii) by selective enrichment and plating on chromogenic agar. While microbial load was generally low, presumptive Cronobacter spp. were detected in five of the edible insect products. Four of the isolates were identified as C. sakazakii, using the Remel RapID ONE biochemical test kit. Genotypic characterisation of the isolates by ITS-PCR, however, demonstrated that the isolates may be other species of Cronobacter instead. Further studies into understanding microbial hazards linked to edible insects for human consumption are required.
Product Description
Millipore
Oxidase Strips, suitable for microbiology