MilliporeSigma
  • 1-(1-Arylethylidene)thiosemicarbazide derivatives: a new class of tyrosinase inhibitors.
1-(1-Arylethylidene)thiosemicarbazide derivatives: a new class of tyrosinase inhibitors.
Bioorganic & medicinal chemistry (2008-03-08)
Jinbing Liu, Wei Yi, Yiqian Wan, Lin Ma, Huacan Song
ABSTRACT
A series of 1-(1-arylethylidene)thiosemicarbazide compounds and their analogues were synthesized and characterized by 1H NMR, MS. Their tyrosinase inhibitory activities were investigated by an assay based on the catalyzing ability of tyrosinase for the oxidation of L-DOPA, comparing with 4-methoxycinnamic acid and arbutin. The results showed that (1) all the synthesized compounds could perform a significant inhibitory activity for tyrosinase; (2) for these compounds, the main active moiety interacting with the center of tyrosinase would be thiosemicarbazo group; (3) the inhibitory activity was close related with thiosemicarbazide moieties and the groups attached on the aromatic ring.
Sigma-Aldrich
Acetanisole, ≥98%, FCC, FG
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%
Sigma-Aldrich
Thiosemicarbazide, 99%
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
4′-Methoxyacetophenone, 99%
Sigma-Aldrich
Tropolone, 98%