MilliporeSigma
  • Canine umbilical cord perivascular tissue: A source of stem cells for therapy and research.
Canine umbilical cord perivascular tissue: A source of stem cells for therapy and research.
Research in veterinary science (2020-02-23)
Rodrigo Viana Sepúlveda, Renato Barros Eleotério In Memorian, Fabrício Luciani Valente, Fabiana Rocha Araújo, Adriano de Paula Sabino, Fernanda Cristina Gontijo Evangelista, Emily Correna Carlo Reis, Andréa Pacheco Batista Borges
ABSTRACT
There are numerous sources of multipotent mesenchymal stromal cells (MSC) with therapeutic potential, and bone marrow is the main one. However, pain, lack of donors and comorbidities associated with harvesting stimulate the search for new sources of MSCs. The aim of this work is to obtain cells from umbilical cord (UC) perivascular tissue of dogs and characterize them as MSCs. For this, the UC was obtained from therapeutic cesarean sections and submitted to enzymatic digestion. The obtained cells were subjected to growth and proliferation tests, as well as the analysis of surface markers, differentiation test in three mesenchymal lineages and analysis of differentiation markers expression. From all the UC used in this study an adherent with fibroblastoid shape cell was obtained, with an initial number of 4.8 × 105 of cells. The growth curves showed a lag phase from 0 to 24 h, followed by a phase of growth of 24 to 168 h, and then phase of cell decay. The doubling time was kept around 15 h until the sixth passage, from which there were signs of cellular senescence. The differentiation assays demonstrated the ability of cells to differentiate into osteoblasts, adipocytes and chondrocytes when subjected to the induction mediums. The study of surface markers was positive for adhesion markers and negative for hematopoietic markers. Thus, cells obtained from canine UC perivascular tissue by enzymatic digestion are multipotent MSC and the protocol developed ensures the perivascular origin of these cells.
Sigma-Aldrich
Phosphate Buffered Saline with 3% Non-Fat Milk, pH 7.4