MilliporeSigma
  • Synthesis and free radical scavenging activity of some new spiropyranocoumarins.
Synthesis and free radical scavenging activity of some new spiropyranocoumarins.
Bioorganic & medicinal chemistry letters (2008-10-07)
Vassiliki Panteleon, Ioannis K Kostakis, Panagiotis Marakos, Nicole Pouli, Ioanna Andreadou
ABSTRACT
A series of novel spiro-substituted 4-hydroxypyranocoumarins and their corresponding dihydropyrano cis-diols has been synthesized. Among them the spiroadamantylpyranocoumarin and the diols can interact with the stable free radical 1,1-diphenyl-2-picrylhydrazyl and scavenge superoxide anions generated in the xanthine-xanthine oxidase system.
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
4-Hydroxycoumarin, 98%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
Supelco
3,5-Di-tert-butyl-4-hydroxytoluene, analytical standard