MilliporeSigma
  • 5-Fluorouracil mediated anti-cancer activity in colon cancer cells is through the induction of Adenomatous Polyposis Coli: Implication of the long-patch base excision repair pathway.
5-Fluorouracil mediated anti-cancer activity in colon cancer cells is through the induction of Adenomatous Polyposis Coli: Implication of the long-patch base excision repair pathway.
DNA repair (2014-12-03)
Dipon Das, Ranjan Preet, Purusottam Mohapatra, Shakti Ranjan Satapathy, Sumit Siddharth, Tigist Tamir, Vaibhav Jain, Prasad V Bharatam, Michael D Wyatt, Chanakya Nath Kundu
ABSTRACT
Colorectal cancer (CRC) patients with APC mutations do not benefit from 5-FU therapy. It was reported that APC physically interacts with POLβ and FEN1, thus blocking LP-BER via APC's DNA repair inhibitory (DRI) domain in vitro. The aim of this study was to elucidate how APC status affects BER and the response of CRC to 5-FU. HCT-116, HT-29, and LOVO cells varying in APC status were treated with 5-FU to evaluate expression, repair, and survival responses. HCT-116 expresses wild-type APC; HT-29 expresses an APC mutant that contains DRI domain; LOVO expresses an APC mutant lacking DRI domain. 5-FU increased the expression of APC and decreased the expression of FEN1 in HCT-116 and HT-29 cells, which were sensitized to 5-FU when compared to LOVO cells. Knockdown of APC in HCT-116 rendered cells resistant to 5-FU, and FEN1 levels remained unchanged. Re-expression of full-length APC in LOVO cells caused sensitivity to 5-FU, and decreased expression of FEN1. These knockdown and addback studies confirmed that the DRI domain is necessary for the APC-mediated reduction in LP-BER and 5-FU. Modelling studies showed that 5-FU can interact with the DRI domain of APC via hydrogen bonding and hydrophobic interactions. 5-FU resistance in CRC occurs with mutations in APC that disrupt or eliminate the DRI domain's interaction with LP-BER. Understanding the type of APC mutation should better predict 5-FU resistance in CRC than simply characterizing APC status as wild-type or mutant.
Sigma-Aldrich
Anti-APC (Ab-1) Mouse mAb (FE9), liquid, clone FE9, Calbiochem®