MilliporeSigma
  • Binding of Amyloid β(1-42)-Calmodulin Complexes to Plasma Membrane Lipid Rafts in Cerebellar Granule Neurons Alters Resting Cytosolic Calcium Homeostasis.
Binding of Amyloid β(1-42)-Calmodulin Complexes to Plasma Membrane Lipid Rafts in Cerebellar Granule Neurons Alters Resting Cytosolic Calcium Homeostasis.
International journal of molecular sciences (2021-03-07)
Joana Poejo, Jairo Salazar, Ana M Mata, Carlos Gutierrez-Merino
ABSTRACT
Lipid rafts are a primary target in studies of amyloid β (Aβ) cytotoxicity in neurons. Exogenous Aβ peptides bind to lipid rafts, which in turn play a key role in Aβ uptake, leading to the formation of neurotoxic intracellular Aβ aggregates. On the other hand, dysregulation of intracellular calcium homeostasis in neurons has been observed in Alzheimer's disease (AD). In a previous work, we showed that Aβ(1-42), the prevalent Aβ peptide found in the amyloid plaques of AD patients, binds with high affinity to purified calmodulin (CaM), with a dissociation constant ≈1 nM. In this work, to experimentally assess the Aβ(1-42) binding capacity to intracellular CaM, we used primary cultures of mature cerebellar granule neurons (CGN) as a neuronal model. Our results showed a large complexation of submicromolar concentrations of Aβ(1-42) dimers by CaM in CGN, up to 120 ± 13 picomoles of Aβ(1-42) /2.5 × 106 cells. Using fluorescence microscopy imaging, we showed an extensive co-localization of CaM and Aβ(1-42) in lipid rafts in CGN stained with up to 100 picomoles of Aβ(1-42)-HiLyteTM-Fluor555 monomers. Intracellular Aβ(1-42) concentration in this range was achieved by 2 h incubation of CGN with 2 μM Aβ(1-42), and this treatment lowered the resting cytosolic calcium of mature CGN in partially depolarizing 25 mM potassium medium. We conclude that the primary cause of the resting cytosolic calcium decrease is the inhibition of L-type calcium channels of CGN by Aβ(1-42) dimers, whose activity is inhibited by CaM:Aβ(1-42) complexes bound to lipid rafts.
Sigma-Aldrich
Anti-β-Amyloid antibody, Mouse monoclonal, clone NAB 228, purified from hybridoma cell culture