MilliporeSigma
  • Physicochemical study and characterization of the trimethoprim/2-hydroxypropyl-γ-cyclodextrin inclusion complex.
Physicochemical study and characterization of the trimethoprim/2-hydroxypropyl-γ-cyclodextrin inclusion complex.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-11-08)
Osmir F L Macedo, George R S Andrade, Leila S Conegero, Ledjane S Barreto, Nivan B Costa, Iara F Gimenez, Luis E Almeida, Daniela Kubota
ABSTRACT
Here we report the preparation of a trimethoprim/2-hydroxypropyl-γ-cyclodextrin inclusion complex along with a physicochemical study, structural characterization, and molecular modeling of the complex. As main results, we observed from phase-solubility studies at two temperatures (20 °C and 35 °C) that the association constants decrease with increasing temperature. Values for K(1:1) constant were of the same magnitude order of those found for the parent γ-CD. The inclusion orientation as evidenced by ROESY measurements involves the inclusion of the 3,4,5-trimethoxybenzyl ring in the CD cavity from the larger rim. This is in agreement with semiempirical molecular modeling calculation.
Sigma-Aldrich
(2-Hydroxypropyl)-γ-cyclodextrin, solid