MilliporeSigma
  • MyD88 deficiency ameliorates β-amyloidosis in an animal model of Alzheimer's disease.
MyD88 deficiency ameliorates β-amyloidosis in an animal model of Alzheimer's disease.
The American journal of pathology (2011-07-19)
Jeong-Eun Lim, Jinghong Kou, Min Song, Abhinandan Pattanayak, Jingji Jin, Robert Lalonde, Ken-ichiro Fukuchi
ABSTRACT
The accumulation of β-amyloid protein (Aβ) in the brain is thought to be a primary etiologic event in Alzheimer's disease (AD). Fibrillar Aβ plaques, a hallmark of AD abnormality, are closely associated with activated microglia. Activated microglia have contradictory roles in the pathogenesis of AD, being either neuroprotective (by clearing harmful Aβ and repairing damaged tissues) or neurotoxic (by producing proinflammatory cytokines and reactive oxygen species). Aβ aggregates can activate microglia by interacting with multiple toll-like receptors (TLRs), the pattern-recognition receptors of the innate immune system. Because the adapter protein MyD88 is essential for the downstream signaling of all TLRs, except TLR3, we investigated the effects of MyD88 deficiency (MyD88(-/-)) on Aβ accumulation and microglial activation in an AD mouse model. MyD88 deficiency decreased Aβ load and microglial activation in the brain. The decrease in Aβ load in an MyD88(-/-) AD mouse model was associated with increased and decreased protein expression of apolipoprotein E (apoE) and CX3CR1, respectively, compared with that in an MyD88 wild-type AD mouse model. These results suggest that MyD88 deficiency may reduce Aβ load by enhancing the phagocytic capability of microglia through fractalkine (the ligand of CX3CR1) signaling and by promoting apoE-mediated clearance of Aβ from the brain. These findings also suggest that chronic inflammatory responses induced by Aβ accumulation via the MyD88-dependent signaling pathway exacerbate β-amyloidosis in AD.
Sigma-Aldrich
Anti-IDE, N-Terminal (97-273) Rabbit pAb, liquid, Calbiochem®