MilliporeSigma
  • Tau-induced upregulation of C/EBPβ-TRPC1-SOCE signaling aggravates tauopathies: A vicious cycle in Alzheimer neurodegeneration.
Tau-induced upregulation of C/EBPβ-TRPC1-SOCE signaling aggravates tauopathies: A vicious cycle in Alzheimer neurodegeneration.
Aging cell (2020-08-21)
Jinwang Ye, Ying Yin, Yaling Yin, Huaqiu Zhang, Huali Wan, Lu Wang, Yue Zuo, Di Gao, Mengzhu Li, Jun Li, Yanchao Liu, Dan Ke, Jian-Zhi Wang
ABSTRACT
Intracellular accumulating of the hyperphosphorylated tau plays a pivotal role in neurodegeneration of Alzheimer disease (AD), but the mechanisms underlying the gradually aggravated tau hyperphosphorylation remain elusive. Here, we show that increasing intracellular tau could upregulate mRNA and protein levels of TRPC1 (transient receptor potential channel 1) with an activated store-operated calcium entry (SOCE), an increased intraneuronal steady-state [Ca2+ ]i , an enhanced endoplasmic reticulum (ER) stress, an imbalanced protein kinases and phosphatase, and an aggravated tauopathy. Furthermore, overexpressing TRPC1 induced ER stress, kinases-phosphatase imbalance, tau hyperphosphorylation and cognitive deficits in cultured neurons and mice, while pharmacological inhibiting or knockout TRPC1 attenuated the hTau-induced deregulations in SOCE, ER homeostasis, kinases-phosphatase balance, and tau phosphorylation level with improved synaptic and cognitive functions. Finally, an increased CCAAT-enhancer-binding protein (C/EBPβ) activity was observed in hTau-overexpressing cells and the hippocampus of the AD patients, while downregulating C/EBPβ by siRNA abolished the hTau-induced TRPC1 upregulation. These data reveal that increasing intracellular tau can upregulate C/EBPβ-TRPC1-SOCE signaling and thus disrupt phosphorylating system, which together aggravates tau pathologies leading to a chronic neurodegeneration.
Sigma-Aldrich
Anti-TRPC1 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Pricing and availability is not currently available.