MilliporeSigma
  • Genome-wide CRISPR-Cas9 Screen Identifies Leukemia-Specific Dependence on a Pre-mRNA Metabolic Pathway Regulated by DCPS.
Genome-wide CRISPR-Cas9 Screen Identifies Leukemia-Specific Dependence on a Pre-mRNA Metabolic Pathway Regulated by DCPS.
Cancer cell (2018-02-27)
Takuji Yamauchi, Takeshi Masuda, Matthew C Canver, Michael Seiler, Yuichiro Semba, Mohammad Shboul, Mohammed Al-Raqad, Manami Maeda, Vivien A C Schoonenberg, Mitchel A Cole, Claudio Macias-Trevino, Yuichi Ishikawa, Qiuming Yao, Michitaka Nakano, Fumio Arai, Stuart H Orkin, Bruno Reversade, Silvia Buonamici, Luca Pinello, Koichi Akashi, Daniel E Bauer, Takahiro Maeda
ABSTRACT
To identify novel targets for acute myeloid leukemia (AML) therapy, we performed genome-wide CRISPR-Cas9 screening using AML cell lines, followed by a second screen in vivo. Here, we show that the mRNA decapping enzyme scavenger (DCPS) gene is essential for AML cell survival. The DCPS enzyme interacted with components of pre-mRNA metabolic pathways, including spliceosomes, as revealed by mass spectrometry. RG3039, a DCPS inhibitor originally developed to treat spinal muscular atrophy, exhibited anti-leukemic activity via inducing pre-mRNA mis-splicing. Humans harboring germline biallelic DCPS loss-of-function mutations do not exhibit aberrant hematologic phenotypes, indicating that DCPS is dispensable for human hematopoiesis. Our findings shed light on a pre-mRNA metabolic pathway and identify DCPS as a target for AML therapy.
Sigma-Aldrich
RG3039, ≥98% (HPLC)