MilliporeSigma
  • Prothymosin alpha is processed to thymosin alpha 1 and thymosin alpha 11 by a lysosomal asparaginyl endopeptidase.
Prothymosin alpha is processed to thymosin alpha 1 and thymosin alpha 11 by a lysosomal asparaginyl endopeptidase.
The Journal of biological chemistry (2003-01-30)
Concepción S Sarandeses, Guillermo Covelo, Cristina Díaz-Jullien, Manuel Freire
ABSTRACT
Thymosin alpha(1) (T alpha(1)) and thymosin T alpha(11) (T alpha(11)) are polypeptides with immunoregulatory properties first isolated from thymic extracts, corresponding to the first 28 and 35 amino acid residues, respectively, of prothymosin alpha (ProT alpha), a protein involved in chromatin remodeling. It has been widely supposed that these polypeptides are not natural products of the in vivo processing of ProT alpha, since neither was found in extracts in which proteolysis was prevented. Here we show that a lysosomal asparaginyl endopeptidase is able to process ProT alpha to generate T alpha(1) and T alpha(11). In view of its catalytic properties and structural and immunological analyses, this protease was identified as mammalian legumain. It selectively cleaves some of the asparaginyl-glycine residues in the ProT alpha sequence; specifically, Asn(28)-Gly(29) and Asn(35)-Gly(36) residues are cleaved with similar efficiency in vitro to generate T alpha(1) and T alpha(11), respectively. By contrast T alpha(1) is the main product detected in vivo, free in the cytosol, at concentrations similar to that of ProT alpha. The data here reported demonstrate that T alpha(1) is not an artifact but rather is naturally present in diverse mammalian tissues and raise the possibility that it has a functional role.
Sigma-Aldrich
Thymosin α1 bovine, ≥90% (HPLC)