MilliporeSigma
  • A practical synthesis of kifunensine analogues as inhibitors of endoplasmic reticulum alpha-mannosidase I.
A practical synthesis of kifunensine analogues as inhibitors of endoplasmic reticulum alpha-mannosidase I.
The Journal of organic chemistry (2005-11-19)
Kirk W Hering, Khanita Karaveg, Kelley W Moremen, William H Pearson
ABSTRACT
[reaction: see text] A practical synthesis of the potent class I alpha-mannosidase inhibitor kifunensine (1) beginning from the inexpensive and readily available starting material L-ascorbic acid (15) is described. The protected amino-alcohol ((2R,3R,4R,5R)-5-amino-2,3:4,6-diisopropylidenedioxyhexanol, 11) served as a key intermediate from which several N-1 substituted kifunensine analogues (including N-methyl, N-cyclohexyl, and N-bis(hydroxymethyl)methyl) and 2-desoxakifunensine analogues (including N-H and N-methyl) were prepared and screened for inhibition of human endoplasmic reticulum alpha-mannosidase I (ER Man I) and mouse Golgi alpha-mannosidase IA (Golgi Man IA). In addition, several pseudodisaccharide kifunensine analogues in which a mannose residue was tethered to N-1 of kifunensine via a two-, three-, or four-carbon linker and an affinity-bound kifunensine analogue were also prepared and evaluated for biological activity. While the synthesized N-1 kifunesine analogues were found to be less potent inhibitors of Class I alpha-mannosidases than kifuensine itself, the bis(hydroxymethyl)methylkifunensine analogue 6 was shown to selectively inhibit ER Man I over Golgi Man IA.
Sigma-Aldrich
Kifunensine, mannosidase inhibitor