MilliporeSigma
  • IDH1 gene mutation activates Smad signaling molecules to regulate the expression levels of cell cycle and biological rhythm genes in human glioma U87‑MG cells.
IDH1 gene mutation activates Smad signaling molecules to regulate the expression levels of cell cycle and biological rhythm genes in human glioma U87‑MG cells.
Molecular medicine reports (2021-03-25)
Yongying Gao, Yanwei Wu, Ningmei Zhang, Hongmei Yuan, Fei Wang, Hui Xu, Jiaxiang Yu, Jie Ma, Shaozhang Hou, Xiangmei Cao
ABSTRACT
Isocitrate dehydrogenase1 (IDH1) mutation is the most important genetic change in glioma. The most common IDH1 mutation results in the amino acid substitution of arginine 132 (Arg/R132), which is located at the active site of the enzyme. IDH1 Arg132His (R132H) mutation can reduce the proliferative rate of glioma cells. Numerous diseases follow circadian rhythms, and there is growing evidence that circadian disruption may be a risk factor for cancer in humans. Dysregulation of the circadian clock serves an important role in the development of malignant tumors, including glioma. Brain‑Muscle Arnt‑Like protein 1 (BMAL1) and Circadian Locomotor Output Cycles Kaput (CLOCK) are the main biological rhythm genes. The present study aimed to further study whether there is an association between IDH1 R132H mutation and biological rhythm in glioma, and whether this affects the occurrence of glioma. The Cancer Genome Atlas (TCGA) database was used to detect the expression levels of the biological rhythm genes BMAL1 and CLOCK in various types of tumor. Additionally, U87‑MG cells were infected with wild‑type and mutant IDH1 lentiviruses. Colony formation experiments were used to detect cell proliferation in each group, cell cycle distribution was detected by flow cytometry and western blotting was used to detect the expression levels of wild‑type and mutant IDH1, cyclins, biological rhythm genes and Smad signaling pathway‑associated genes in U87‑MG cells. TCGA database results suggested that BMAL1 and CLOCK were abnormally expressed in glioma. Cells were successfully infected with wild‑type and mutant IDH1 lentiviruses. Colony formation assay revealed decreased cell proliferation in the IDH1 R132H mutant group. The cell cycle distribution detected by flow cytometry indicated that IDH1 gene mutation increased the G1 phase ratio and decreased the S phase ratio in U87‑MG cells. The western blotting results demonstrated that IDH1 R132H mutation decreased the expression levels of the S phase‑associated proteins Cyclin A and CDK2, and increased the expression levels of the G1 phase‑associated proteins Cyclin D3 and CDK4, but did not significantly change the expression levels of the G2/M phase‑associated protein Cyclin B1. The expression levels of the positive and negative rhythm regulation genes BMAL1, CLOCK, period (PER s (PER1, 2 and 3) and cryptochrom (CRY)s (CRY1 and 2) were significantly decreased, those of the Smad signaling pathway‑associated genes Smad2, Smad3 and Smad2‑3 were decreased, and those of phosphorylated (p)‑Smad2, p‑Smad3 and Smad4 were increased. Therefore, the present results suggested that the IDH1 R132H mutation may alter the cell cycle and biological rhythm genes in U87‑MG cells through the TGF‑β/Smad signaling pathway.
Sigma-Aldrich
Anti-IDH1 (R132H) antibody, Mouse monoclonal, clone HMab-1, purified from hybridoma cell culture