MilliporeSigma
  • Amyloid β and tau pathology in brains of aged pinniped species (sea lion, seal, and walrus).
Amyloid β and tau pathology in brains of aged pinniped species (sea lion, seal, and walrus).
Acta neuropathologica communications (2021-01-09)
Yuta Takaichi, James K Chambers, Kei Takahashi, Yoshiyuki Soeda, Riki Koike, Etsuko Katsumata, Chiaki Kita, Fuko Matsuda, Makoto Haritani, Akihiko Takashima, Hiroyuki Nakayama, Kazuyuki Uchida
ABSTRACT
Alzheimer's disease (AD) is characterized by the accumulation of amyloid-β (Aβ) as senile plaques and cerebral amyloid angiopathy, and hyperphosphorylated tau (hp-tau) as neurofibrillary tangles in the brain. The AD-related pathology has been reported in several non-human animals, and most animals develop only the Aβ or tau pathology. We herein describe the Aβ and hp-tau pathology in the brains of aged pinniped species (seal, sea lion, and walrus). Molecular analyses revealed that the sequence of pinniped Aβ was identical to that of human Aβ. Histopathological examinations detected argyrophilic plaques composed of Aβ associated with dystrophic neurites in the cerebral cortex of aged pinnipeds. Astrogliosis and microglial infiltration were identified around Aβ plaques. Aβ deposits were observed in the blood vessel walls of the meninges and cerebrum. Pinniped tau protein was physiologically subjected to alternative splicing at exons 2, 3, and 10, and presented as five isoforms: two 3-repeat tau isoforms (1N3R, 2N3R) and three 4-repeat tau isoforms (0N4R, 1N4R, 2N4R); 0N3R tau isoform was absent. Histopathological examinations revealed argyrophilic fibrillar aggregates composed of hp-tau in the neuronal somata and neurites of aged pinniped brains. Few hp-tau aggregates were found in oligodendrocytes and microglia. Biochemically, hp-tau of the 3-repeat and 4-repeat isoforms was detected in brain sarkosyl-insoluble fractions. Aβ and hp-tau both predominantly accumulated in the neocortex, particularly the frontal cortex. Furthermore, the activation of GSK-3β was detected within cells containing hp-tau aggregates, and activated GSK-3β was strongly expressed in cases with severe hp-tau pathologies. The present results suggest that, in association with Aβ deposition, the activation of GSK-3β contributes to hp-tau accumulation in pinniped brains. Here, we report that pinniped species naturally accumulate Aβ and tau with aging, similar to the human AD pathology.
Sigma-Aldrich
Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6, culture supernatant, clone 1E1/A6, Upstate®
Sigma-Aldrich
Anti-Amyloid Beta (ABeta) x-42 Antibody, clone 12F4, clone 12F4, from mouse
Sigma-Aldrich
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®