MilliporeSigma
  • Extensive tissue-specific expression variation and novel regulators underlying circadian behavior.
Extensive tissue-specific expression variation and novel regulators underlying circadian behavior.
Science advances (2021-01-31)
Maria Litovchenko, Antonio C A Meireles-Filho, Michael V Frochaux, Roel P J Bevers, Alessio Prunotto, Ane Martin Anduaga, Brian Hollis, Vincent Gardeux, Virginie S Braman, Julie M C Russeil, Sebastian Kadener, Matteo Dal Peraro, Bart Deplancke
ABSTRACT
Natural genetic variation affects circadian rhythms across the evolutionary tree, but the underlying molecular mechanisms are poorly understood. We investigated population-level, molecular circadian clock variation by generating >700 tissue-specific transcriptomes of Drosophila melanogaster (w1118 ) and 141 Drosophila Genetic Reference Panel (DGRP) lines. This comprehensive circadian gene expression atlas contains >1700 cycling genes including previously unknown central circadian clock components and tissue-specific regulators. Furthermore, >30% of DGRP lines exhibited aberrant circadian gene expression, revealing abundant genetic variation-mediated, intertissue circadian expression desynchrony. Genetic analysis of one line with the strongest deviating circadian expression uncovered a novel cry mutation that, as shown by protein structural modeling and brain immunohistochemistry, disrupts the light-driven flavin adenine dinucleotide cofactor photoreduction, providing in vivo support for the importance of this conserved photoentrainment mechanism. Together, our study revealed pervasive tissue-specific circadian expression variation with genetic variants acting upon tissue-specific regulatory networks to generate local gene expression oscillations.
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Propionic acid, ≥99.5%