MilliporeSigma
Feedback regulation of insulin secretion by extended synaptotagmin-1.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-12-28)
Beichen Xie, Phuoc My Nguyen, Olof Idevall-Hagren
ABSTRACT
Endoplasmic reticulum (ER)-plasma membrane (PM) contacts are dynamic structures with important roles in the regulation of calcium (Ca2+) and lipid homeostasis. The extended synaptotagmins (E-Syts) are ER-localized lipid transport proteins that interact with PM phosphatidylinositol 4,5-bisphosphate in a Ca2+-dependent manner. E-Syts bidirectionally transfer glycerolipids, including diacylglycerol (DAG), between the 2 juxtaposed membranes, but the biologic significance of this transport is still unclear. Using insulin-secreting cells and live-cell imaging, we now show that Ca2+-triggered exocytosis of insulin granules is followed, in sequence, by PM DAG formation and E-Syt1 recruitment. E-Syt1 counteracted the depolarization-induced DAG formation through a mechanism that required both voltage-dependent Ca2+ influx and Ca2+ release from the ER. E-Syt1 knockdown resulted in prolonged accumulation of DAG in the PM, resulting in increased glucose-stimulated insulin secretion. We conclude that Ca2+-triggered exocytosis is temporally coupled to Ca2+-triggered E-Syt1 PM recruitment and removal of DAG to negatively regulate the same process.-Xie, B., Nguyen, P. M., Idevall-Hagren, O. Feedback regulation of insulin secretion by extended synaptotagmin-1.
Sigma-Aldrich
Anti-ESYT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ESYT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-ESYT3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution