MilliporeSigma
  • The protective effects of fibroblast growth factor 10 against hepatic ischemia-reperfusion injury in mice.
The protective effects of fibroblast growth factor 10 against hepatic ischemia-reperfusion injury in mice.
Redox biology (2021-01-15)
Santie Li, Zhongxin Zhu, Mei Xue, Xuebo Pan, Gaozan Tong, Xinchu Yi, Junfu Fan, Yuankuan Li, Wanqian Li, Yetong Dong, Enzhao Shen, Wenjie Gong, Xuejiao Wang, Ying Yu, Yoo Jae Maeng, Xiaokun Li, Kwang Youl Lee, Litai Jin, Weitao Cong
ABSTRACT
Hepatic ischemia-reperfusion injury (IRI) is a major complication of liver surgery and transplantation. IRI leads to hepatic parenchymal cell death, resulting in liver failure, and lacks effective therapeutic approaches. Fibroblast growth factor 10 (FGF10) is a paracrine factor which is well-characterized with respect to its pro-proliferative effects during embryonic liver development and liver regeneration, but its role in hepatic IRI remains unknown. In this study, we investigated the role of FGF10 in liver IRI and identified signaling pathways regulated by FGF10. In a mouse model of warm liver IRI, FGF10 was highly expressed during the reperfusion phase. In vitro experiments demonstrated that FGF10 was primarily secreted by hepatic stellate cells and acted on hepatocytes. The role of FGF10 in liver IRI was further examined using adeno-associated virus-mediated gene silencing and overexpression. Overexpression of FGF10 alleviated liver dysfunction, reduced necrosis and inflammation, and protected hepatocytes from apoptosis in the early acute injury phase of IRI. Furthermore, in the late phase of IRI, FGF10 overexpression also promoted hepatocyte proliferation. Meanwhile, gene silencing of FGF10 had the opposite effect. Further studies revealed that overexpression of FGF10 activated nuclear factor-erythroid 2-related factor 2 (NRF2) and decreased oxidative stress, mainly through activation of the phosphatidylinositol-3-kinase/AKT pathway, and the protective effects of FGF10 overexpression were largely abrogated in NRF2 knockout mice. These results demonstrate the protective effects of FGF10 in liver IRI, and reveal the important role of NRF2 in FGF10-mediated hepatic protection during IRI.
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR2
Sigma-Aldrich
Anti-FGF10 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
MISSION® esiRNA, targeting human FGFR1 (1)
Sigma-Aldrich
MISSION® esiRNA, targeting human FGF10